Tisk…

2,6-Difluoro-3-methylbenzylamine 97%

Dodavatel: Thermo Scientific

Nebezpečí

H26506.06EA 10400 CZK
H26506.06 H26506.03
2,6-Difluoro-3-methylbenzylamine 97%
2,6-Difluoro-3-methylbenzylamine
Vzorec: C₈H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD01631410
CAS číslo: 261763-42-2
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR