Tisk…

4-(Methylthio)phenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL550426-25GEA 0 CZK
SIAL550426-25G
4-(Methylthio)phenol, Sigma-Aldrich®
4-(Methylthio)phenol
Vzorec: C₇H₈OS
MW: 140,21
MDL Number: MFCD00002351
CAS číslo: 1073-72-9
EINECS: 214-031-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR