2-Mercapto-5-methoxybenzothiazole ≥97%
L14228.03EA
1390
CZK
L14228.03
2-Mercapto-5-methoxybenzothiazole ≥97%
2-Mercapto-5-methoxybenzothiazole
Vzorec:
C₈H₇NOS₂ MW: 197,28 g/mol Bod tání: 193…198 °C |
MDL Number:
MFCD00185941 CAS číslo: 55690-60-3 EINECS: 259-755-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...