Tisk…

4-(Trifluoromethylthio)benzylamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL555460-1GEA 7530 CZK
SIAL555460-1G
4-(Trifluoromethylthio)benzylamine, Sigma-Aldrich®
4-(Trifluoromethylthio)benzylamine
Vzorec: C₈H₈F₃NS
MW: 207,21 g/mol
MDL Number: MFCD00190129
CAS číslo: 128273-56-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR