Purine ≥97%
Dodavatel: Thermo Scientific
J60999.03EA
5980
CZK
J60999.03
Purine ≥97%
Purine
Vzorec:
C₅H₄N₄ MW: 120,11 g/mol Bod varu: 265 °C (1013 hPa) Bod tání: 215…217 °C Hustota: 1,22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00079221 CAS číslo: 120-73-0 EINECS: 204-421-2 Merck Index: 12,08129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...