Sodium cholate
Dodavatel: Thermo Scientific
Varování
|
J62050.30EA
8250
CZK
J62050.30
J62050.22
J62050.14
Sodium cholate
Sodium cholate
Biochemical solubilizing agent
Vzorec:
C₂₄H₃₉NaO₅ MW: 430,56 g/mol Bod tání: 198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150749 CAS číslo: 361-09-1 EINECS: 206-643-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...