Tisk…
AFFY75896
USBM75896100MLEA 941 CZK
USBM75896100ML
AFFY75896
2M solution of potassium chloride (KCl) in high purity dH2O. Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles.
Contaminant Testing:
DNase (endo): Passes test
DNase (exo): Passes test
RNase: Passes test