Tisk…

ALFAJ60688

J60688.AKEA 1970 CZK
ALFAJ60688.AK ALFAJ60688.AP
ALFAJ60688
CAS No.: 631-61-8
Molecular Formula: C2H7NO2
Formula Weight: 77.08
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00013066
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR