Tisk…

4-(Trifluoromethoxy)benzamidoxime, 97%

H58879.06EA 5530 CZK
ALFAH58879.06 ALFAH58879.03
4-(Trifluoromethoxy)benzamidoxime, 97%
Purity: 97%
CAS No.: 56935-71-8
Molecular Formula: C8H7F3N2O2
Formula Weight: 220.15
Melting Point: 113-115°
MDL No.: MFCD00085175
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR