(S)-(+)-Ethyl-3-hydroxybutyrate
Dodavatel: Thermo Scientific
Varování
|
J63184.03EA
2530
CZK
J63184.03
J63184.06
(S)-(+)-Ethyl-3-hydroxybutyrate
(S)-(+)-Ethyl-3-hydroxybutyrate
For synthesis of optically active products
Vzorec:
C₆H₁₂O₃ MW: 132,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066206 CAS číslo: 56816-01-4 EINECS: 260-393-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...