Tisk…

Fenvalerate ≥99%

Dodavatel: Thermo Scientific

J63562.MCEA 9510 CZK
J63562.MC
Fenvalerate ≥99%
Fenvalerate
Vzorec: C₂₅H₂₂ClNO₃
MDL Number: MFCD00055324
CAS číslo: 51630-58-1
EINECS: 257-326-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR