Tisk…

BigCHAP

J64631.06EA 12450 CZK
ALFAJ64631.06 ALFAJ64631.MD ALFAJ64631.03
BigCHAP
CAS No.: 86303-22-2
Molecular Formula: C42H75N3O16
Formula Weight: 878.07
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 136-145°
MDL No.: MFCD00082541

Non-ionic detergent for anion exchange chromatography
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR