Tisk…
Spill sump, PIG®
Dodavatel: New Pig
PIG®
NEWPPAK360EA 2730 CZK
NEWPPAK360 NEWPPAK361
Spill sump, PIG®
Skladování chemikálií
These stackable spill sumps are ideal to catch leaks and drips before they hit the floor.
  • Made of linear LDPE with inhibitors
  • Rust- and corrosion-free
  • Sumps can be double stacked with drums in place to save storage space
  • Compatible with a wide variety of chemicals