Tisk…
VWR® Protease Inhibitor Cocktail, Bacterial
Dodavatel: Avantor
VWR®
M306-5MLEA 6390 CZK
M306-5ML
VWR® Protease Inhibitor Cocktail, Bacterial
Doplňky a přísady médií z buněčných kultur
Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, EDTA and Pepstatin. Each vial contains sufficient material to provide 5 ml of a 20X solution upon reconstitution.