Order Entry
Ion exchange chromatography media, DEAE Sepharose™ Fast Flow
Ion exchange chromatography media, DEAE Sepharose™ Fast Flow
Supplier:  Cytiva
Certificates
undefined
Ion exchange chromatography media, DEAE Sepharose™ Fast Flow
Supplier:  Cytiva
Certificates
Specifications
  • pH range:
    2 - 12
  • Matrix:
    Cross-linked agarose, 6%, spherical
  • Ligand:
    Diethylaminoethyl
  • Storage temperature:
    4 - 30 °C
  • Total ionic capacity:
    0,12 - 0,15 mmol (Cl-)/ml
Specifications
Product Information
  • Catalog #
    Supplier #
    Type
    Particle Size
    Exclusion limit
    Pk
    Availability
    Price
  • DEAE Sepharose™ Fast Flow
    DEAE Sepharose™ Fast Flow
    17-0709-10
    17-0709-10
    Liquid
    ~90 µm
    ~4 × 106 globular proteins
    25 mL
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 2 980,00 Kč
    2 980,00 Kč
    1 * 25 mL
       2 980,00 Kč
  • DEAE Sepharose™ Fast Flow
    DEAE Sepharose™ Fast Flow
    17-0709-01
    17-0709-01
    Liquid
    ~90 µm
    ~4 × 106 globular proteins
    500 ml
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 23 110,00 Kč
    23 110,00 Kč
    1 * 500 mL
       23 110,00 Kč