Order Entry
Cation exchange chromatography columns, HiTrap™ SP FF
Cation exchange chromatography columns, HiTrap™ SP FF
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap™ SP FF
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-5054-01
  • 17-5157-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 4 - 13
  • 4 - 13
Specifications
Product Information
  • Catalog #
    Supplier #
    Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Flow rate
    Availability
    Price
  • HisTrap™ SP FF
    HisTrap™ SP FF
    17-5054-01
    17-5054-01
    PP
    90 µm
    7×25 mm
    1 ml
    <4 ml/min
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 5 270,00 Kč
    5 270,00 Kč
    1 * 5 KS
       5 270,00 Kč
    Supplier Article Number: 17-5054-01
    Local Article Number: GEHE17-5054-01
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Max. pressure
    5 bar
    pH range
    4 - 13
  • HisTrap™ SP FF
    HisTrap™ SP FF
    17-5157-01
    17-5157-01
    PP
    90 µm
    16×25 mm
    5 ml
    <20 ml/min
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 11 460,00 Kč
    11 460,00 Kč
    1 * 5 KS
       11 460,00 Kč
    Supplier Article Number: 17-5157-01
    Local Article Number: GEHE17-5157-01
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Max. pressure
    5 bar
    pH range
    4 - 13