Tisk…
Precision balances, Practum® series
Dodavatel: Sartorius
611-2659EA 54850 CZK
611-2659 611-2658 611-2656 611-2895 611-2664 611-2894 611-2663 611-2893 611-2662 611-2661 611-2892 611-2891 611-2890
Precision balances, Practum® series
Váhy
A practical basic balance that incorporates Sartorius' renowned weighing technology to deliver reliable professional results. Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, and averaging. Results can be transferred via a mini-USB interface to a printer or a computer directly into Microsoft Excel® spreadsheet without additional software. Menu with choice of 10 languages.
  • High contrast colour touch screen
  • Automatic calibration and adjustment possible with an optional external weight
  • Robust housing design and rugged weigh cell
  • Supervisor lock to prevent any unintentional changes to settings
  • Built-in overload protection
Informace o dodávce: Supplied with universal power adapter, in-use cover and application guide but without calibration weight.