Tisk…
DNA silver staining kit, PlusOne®
Dodavatel: Cytiva
17-6000-30EA 11720 CZK
GEHE17-6000-30
DNA silver staining kit, PlusOne®
The DNA silver staining kit brings speed, convenience and very high sensitivity to the staining of proteins and nucleic acids.
  • Visualization technique to detect nucleic acids in the picogram range with a colourless background
  • Typical sensitivity is 20 to 50 pg DNA/band
  • Minimized production of toxic waste and optimised sensitivity and robustness for automatic staining equipment, such as Processor Plus
Provides reproducible and sensitive nonradioactive detection of nucleic acids in polyacrylamide gels in less than 90 minutes.
Informace pro objednávání: Includes protocols, fixing solution, staining solution, sodium carbonate solution, 37% formaldehyde, 2% sodium thiosulfate, stopping and preserving solution.