Tisk…
DNA marker, KiloBase™
Dodavatel: Cytiva
27-4004-01EA 8570 CZK
GEHE27-4004-01
DNA marker, KiloBase™
KiloBase™ DNA Marker is used for quick and precise sizing of double-stranded DNA fragments from 500 bp to 10 kb on agarose gels.
  • Supplied at 0,5 µg/µl in TE buffer (pH 8) in sufficient quantity for 100 loadings using 1 µl per loading after mixing with tracking dye
  • Consists of 11 easily-identifiable bands at 0,5, 1, 1,5, 2, 2,5, 3, 4, 5, 6, 8 and 10 kb when run on a 0,8% agarose gel
  • Bands stain with approximately the same intensity, except for the 1 kb band, which is three times the intensity of the other bands
Not designed for quantitating DNA content in a sample.