Tisk…
Needles with SmartSlip™ technology, BD Eclipse™
Dodavatel: BD Medical
BD Eclipse™
BDAM305771EA 1020 CZK
BDAM305771
Needles with SmartSlip™ technology, BD Eclipse™
Jehly ke stříkačkám
  • PP standard hub fits all Luer lock syringes
  • No hard surface required for activation
  • Bevel oriented needle allows for low-angle injections
Wide textured finger pad provides safe activation area for thumb or forefinger.