Tisk…
Affinity chromatography columns, HiTrap™ Streptavidin HP
Dodavatel: Cytiva
HiTrap™
17-5112-01EA 21510 CZK
17-5112-01
Affinity chromatography columns, HiTrap™ Streptavidin HP
Kolony chromatografické
HiTrap Streptavidin HP is a column prepacked with Streptavidin Sepharose High Performance for purification of biotinylated proteins and biomolecules.
  • Convenient, fast, reliable purification of biotinylated compounds
  • The strong interactions of biotin and streptavidin, as well as the somewhat weaker interactions of 2-iminobiotin and streptavidin are utilised
  • The column can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System
Streptavidin Sepharose™ High Performance is also available in lab packs, using the same purified streptavidin isolated from Streptomyces avidinii bound to Sepharose™ High Performance. The prepacked medium ensures reproducibility, high performance, and excellent physical and chemical stability.