Tisk…
Wash buffer, PBS with 0,05% Tween® 20, pH 7,4, VWR®
Dodavatel: VWR Life Science
M245-10.4G-5PKEA 3060 CZK
M245-10.4G-5PK
Wash buffer, PBS with 0,05% Tween® 20, pH 7,4, VWR®
Pufry Pufry pro elektroforézu Pufry promývací
Pre-blended buffer supplied as a ready to use pouch. Each pack prepares 1 litre of 1X PBS/Tween® 20 solution (10 mmol phosphate buffer, 137 mmol sodium chloride, 2,7 mmol potassium chloride, 0,05% Tween® 20) when pouch contents are added to 1 litre of water.
Conductivity@25 °C (10,38 g/l water) is 5,5 to 16,5.
pH@25 °C (10,38 g/l water) is 7,2 to 7,6.