Tisk…
GEHE17-1332-01
Dodavatel: Cytiva
17-1332-01EA 3900 CZK
17-1332-01
GEHE17-1332-01
PlusOne™ Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane, a low toxicity, environmentally safe solvent. It is used to inhibit sticking of polyacrylamide gels, agarose gels and nucleic acids to glass surfaces.