Chcete se opět přihlásit?
Safety canisters
Dodavatel: Rötzmeier
Stainless steel.
  • In-plant use of flammable liquids, such as solvents
  • Explosion-proof due to flame traps (removable) and pressure control valves
  • Taps and metering devices are self-closing
  • High quality and chemically resistant stainless steel (0,8 mm thick) with PTFE gaskets
  • Easy handling due to ergonomic design
  • Models with self-closing taps are equipped with a separate ventilation screw and are suitable for horizontal storage and decanting
Canister shape maximises the usage of limited storage space, e.g. in safety cabinets.
Po přihlášení uvidíte dostupnost zboží na skladě a ceny.
  Zapamatovat si mě na tomto zařízení
OBJEDNÁVKA
Rozbalit vše / Zavřít vše
Objem Popis Bal. VWR katalogové číslo Množství
5 Safety canister with screw cap 1½" 1 119-2045
5 Safety canister with self-closing metering device 1½" 1 119-0172
5 Safety canister with self-closing tap 1½" and sep. ventilation 1 119-2180
10 Safety canister with screw cap 1½" 1 119-2046
10 Safety canister with self-closing metering device 1½" 1 119-0116
10 Safety canister with self-closing tap 1½" and sep. ventilation 1 119-2181
20 Safety canister with screw cap 1½" 1 119-0121
20 Safety canister with self-closing metering device 1½" and sep. ventilation 1 119-2182
20 Safety canister with self-closing tap 1½" and sep. ventilation 1 119-2380
Generovat čárové kódy Formát popisku čárového kódu : 
Vygenerovat PDF katalog
PŘÍSLUŠENSTVÍ
Rozbalit vše / Zavřít vše
Accessories
Popis Určeno pro Bal. VWR katalogové číslo Množství
Self-closing metering device 1½“ for safety canisters and barrels 119-0172, -0116, -2045, -2046, -2370, -2371, -0115 1 119-2391
Screw cap 1½" with pressure control valve 119-0172, -0116, -2380, -2045, -2046, -0121, -2180, -2181, -2182, -2160, -0111, -0115, -0123 1 119-0129
Self-closing tap 1½" 119-2180, -2181, -2182, -0122, -0157, -0156 1 119-2401
Rotable spout 1½“ All Rötzmeier safety canisters and barrels (10 and 25 l) 1 119-0161
Flame trap All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l) 1 119-0169
Generovat čárové kódy Formát popisku čárového kódu : 
Vygenerovat PDF katalog
Enter Details
Select the article for which you are looking for Certificate
Doplňte Lot číslo. Toto pole je povinné. Jestli číslo neznáte, napíšte "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Nebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Katalogové číslo Availability Jednotka Vaše cena Price Per Množství