Tisk…
ESD safety shoes, lace-up, 8698, Ladies All-round
Dodavatel: Uvex
UVEX8698.2/38EA 2250 CZK
UVEX8698.2/38 UVEX8698.2/39 UVEX8698.2/43 UVEX8698.2/42 UVEX8698.2/35 UVEX8698.2/41
ESD safety shoes, lace-up, 8698, Ladies All-round
Laced ESD shoe, designed for a woman’s foot, is manufacured from breathable, water-resistant smooth leather, mesh lining and a single-layer PUR outsole; with padded collar and tongue and reflectors for enhanced wearer safety.
  • Climazone technology for improved comfort
  • Ultra breathable distance mesh lining
  • Removable 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Anti-twist heel cap for greater stability and protection from ankle twisting
  • Aluminium toecap and pierce-resistant, metal-free midsole
  • Fulfils ESD requirements: resistance to ground <35 megaohms
Colour: Black
EN ISO 20345:2011; S3, SRC