4'-Chloro-2,2,2-trifluoroacetophenone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL112860-10GEA
8520
CZK
SIAL112860-10G
4'-Chloro-2,2,2-trifluoroacetophenone, Sigma-Aldrich®
4'-Chloro-2,2,2-trifluoroacetophenone
Vzorec:
C₈H₄ClF₃O MW: 208,5700 g/mol |
MDL Number:
MFCD00018686 CAS číslo: 321-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...