Tisk…

Cyclopentadienylhafnium trichloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL484369-1GEA 0 CZK
SIAL484369-1G
Cyclopentadienylhafnium trichloride, Sigma-Aldrich®
Cyclopentadienylhafnium trichloride
Vzorec: C₅H₅Cl₃Hf
MDL Number: MFCD01073805
CAS číslo: 61906-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR