Tisk…

Propisochlor, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

34056-50MG-REA 0 CZK
34056-50MG-R
Propisochlor, Supelco®
Propisochlor
Vzorec: C₁₅H₂₂ClNO₂
MDL Number: MFCD01632345
CAS číslo: 86763-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR