Tisk…

3-Formyl-4-methoxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL512869-5GEA 0 CZK
SIAL512869-5G
3-Formyl-4-methoxyphenylboronic acid, Sigma-Aldrich®
3-Formyl-4-methoxyphenylboronic acid
Vzorec: C₈H₉BO₄
MW: 179,97
MDL Number: MFCD02093661
CAS číslo: 121124-97-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR