5-Formyl-2-methoxyphenylboronic acid ≥98%
Dodavatel: Thermo Scientific
L19059.03EA
2130
CZK
L19059.03
L19059.MD
5-Formyl-2-methoxyphenylboronic acid ≥98%
5-Formyl-2-methoxyphenylboronic acid
Vzorec:
C₈H₉BO₄ Bod tání: ∼160 °C |
MDL Number:
MFCD01319044 CAS číslo: 127972-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...