N-(3-Acetyl-4-hydroxyphenyl)butyramide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL636878-5GEA
0
CZK
SIAL636878-5G
N-(3-Acetyl-4-hydroxyphenyl)butyramide, Sigma-Aldrich®
N-(3-Acetyl-4-hydroxyphenyl)butyramide
Vzorec:
C₁₂H₁₅NO₃ MW: 221,26 g/mol |
MDL Number:
MFCD00798556 CAS číslo: 40188-45-2 EINECS: 254-831-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...