Tisk…
Di-tert-butyl iminodicarboxylate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL375276-25GEA 0 CZK
SIAL375276-25G SIAL375276-5G
Di-tert-butyl iminodicarboxylate, Sigma-Aldrich®
Di-tert-butyl iminodicarboxylate
Vzorec: C₁₀H₁₉NO₄
MW: 217,27 g/mol
MDL Number: MFCD00043309
CAS číslo: 51779-32-9