Tisk…

Diclofenac

Dodavatel: EHRENSTORFER GMBH

EHERC12537000EA 2250 CZK
EHERC12537000
Diclofenac
Diclofenac
Vzorec: C₁₄H₁₁Cl₂NO₂
CAS číslo: 15307-86-5
EINECS: 239-348-5
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR