Tisk…

Diclofenac sodium salt

Dodavatel: LGC Standards PROMOCHEM

PROOMM0006.00EA 34880 CZK
PROOMM0006.00
Diclofenac sodium salt
Diclofenac sodium salt
Vzorec: C₁₄H₁₀Cl₂NNaO₂
MDL Number: MFCD00082251
CAS číslo: 15307-79-6
EINECS: 239-346-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR