Tisk…

4'-Hydroxydiclofenac, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIALH3661-1VLEA 41880 CZK
SIALH3661-1VL
4'-Hydroxydiclofenac, Sigma-Aldrich®
4'-Hydroxydiclofenac
Vzorec: C₁₄H₁₁Cl₂NO₃
MW: 312,15 g/mol
MDL Number: MFCD01671980
CAS číslo: 64118-84-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR