Tisk…

4-Chlorophenyl chloroformate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL363871-5GEA 0 CZK
SIAL363871-5G
4-Chlorophenyl chloroformate, Sigma-Aldrich®
4-Chlorophenyl chloroformate
Vzorec: C₇H₄Cl₂O₂
MDL Number: MFCD00000638
CAS číslo: 7693-45-0
UN: 3277
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR