Tisk…

[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL555134-1GEA 0 CZK
SIAL555134-1G
[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride, Sigma-Aldrich®
[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
Vzorec: C₃₄H₂₈Cl₂CoFeP₂
MW: 684,23
MDL Number: MFCD03427161
CAS číslo: 67292-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR