Tisk…

Aniline-[ring-¹³C₆] (99% ¹³C )

CAMBCLM714-0.1 CAMBCLM-714-0.25
Aniline-[ring-¹³C₆] (99% ¹³C )
Aniline-[ring-¹³C₆]
Vzorec: C₆H₇N
MDL Number: MFCD00143617
CAS číslo: 100849-37-4
UN: 1547
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR