Tisk…

Aniline-[ring-¹³C₆] (99% ¹³C ), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL485497-250MGEA 22390 CZK
SIAL485497-250MG
Aniline-[ring-¹³C₆] (99% ¹³C ), Sigma-Aldrich®
Aniline-[ring-¹³C₆]
Vzorec: C₆H₇N
MDL Number: MFCD00143617
CAS číslo: 100849-37-4
UN: 1547
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR