Tisk…

Buffer, CAPSO, 0,2M solution, pH 8,5

J60973.AEEA 808 CZK
ALFAJ60973.AE ALFAJ60973.AK
Buffer, CAPSO, 0,2M solution, pH 8,5
CAS No.: 102601-34-3
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00070063
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR