Tisk…

Buffers, CAPSO, 0,2 M solutions

Dodavatel: Thermo Fisher Scientific
J63242.AEEA 1710 CZK
J63242.AE
Buffers, CAPSO, 0,2 M solutions
Pufry Pufry biologické
3-Cyclohexylamino-2-hydroxy-1-propanesulfonic acid buffer solution.

CAS No.: 102601-34-3
Odor: Odorless
MDL No.: MFCD00070063

Upozornení: Store at ambient temperature.
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR