Tisk…

Buffer, potassium acetate, 1 M aq, soln, pH 7,5

Dodavatel: Thermo Fisher Scientific
J62817.AKEA 1700 CZK
J62817.AK J62817.AE
Buffer, potassium acetate, 1 M aq, soln, pH 7,5
Pufry Pufry klonovací
CAS No.: 127-08-2
Molecular formula: C₂H₃KO₂
Formula weight: 98,15
Physical form: liquid
Appearance: colourless
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR