Tisk…

Buffer, bicarbonate, 1M solution, pH 8,5

Dodavatel: Thermo Fisher Scientific
J60092.APEA 2290 CZK
ALFAJ60092.AP ALFAJ60092.AK
Buffer, bicarbonate, 1M solution, pH 8,5
CAS No.: 144-55-8
Physical form: liquid
MDL No.: MFCD00003528
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR