Tisk…

Diosmin

Dodavatel: U.S. Pharmacopeia

USPH1213329EA 37000 CZK
USPH1213329
Diosmin
Diosmin
Vzorec: C₂₈H₃₂O₁₅
MDL Number: MFCD00009772
CAS číslo: 520-27-4
EINECS: 208-289-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR