Hesperidin
MOLEM14809891EA
0
CZK
MOLEM14809891
Hesperidin
Hesperidin
Vzorec:
C₂₈H₃₄O₁₅ MW: 610,56 g/mol |
MDL Number:
MFCD00075663 CAS číslo: 520-26-3 EINECS: 208-288-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...