Tisk…

N(α)-Boc-L-histidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL408433-5GEA 3020 CZK
SIAL408433-5G
N(α)-Boc-L-histidine, Sigma-Aldrich®
N(α)-Boc-L-histidine
Vzorec: C₁₁H₁₇N₃O₄
MW: 255,3 g/mol
MDL Number: MFCD00065576
CAS číslo: 17791-52-5
EINECS: 241-768-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR