D(-)-Luciferin sodium salt monohydrate
MOLE25302670-25MGEA
0
CZK
MOLE25302670-25MG
D(-)-Luciferin sodium salt monohydrate
D-(-)-Luciferin sodium salt monohydrate
Vzorec:
C₁₁H₇N₂NaO₃S₂·H₂O MW: 320,32 g/mol |
MDL Number:
MFCD11865368 CAS číslo: 103404-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...