Tisk…

Hydrochlorothiazide

Dodavatel: LGC Standards PROMOCHEM

PROOICRS0324EA 7050 CZK
PROOICRS0324
Hydrochlorothiazide
Hydrochlorothiazide
Vzorec: C₇H₈ClN₃O₄S₂
MDL Number: MFCD00051765
CAS číslo: 58-93-5
EINECS: 200-403-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR