Ergocalciferol (vitamin D2, calciferol)
Dodavatel: US Biological
USBIE3452EA
6660
CZK
USBIE3452
Ergocalciferol (vitamin D2, calciferol)
Ergocalciferol (vitamin D2, calciferol)
Vzorec:
C₂₈H₄₄O MW: 396,66 g/mol |
MDL Number:
MFCD00166988 CAS číslo: 50-14-6 EINECS: 200-014-9 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...