(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL223859-5GEA
2760
CZK
SIAL223859-5G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide, Sigma-Aldrich®
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
Vzorec:
C₂₂H₂₂BrO₂P MW: 429,30 g/mol |
MDL Number:
MFCD00011966 CAS číslo: 52509-14-5 EINECS: 257-977-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...